Product Details

SNP ID
rs976025748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:17789732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTCGCGTAGGACGCACCAGATG[C/T]CGCTGTGGCTCCCGCACCCGGGCGC
Phenotype
MIM: 608882
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MICAL3 PubMed Links

Gene Details

Gene
MICAL3
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122731.2 7362 Intron NP_001116203.1
NM_001136004.3 7362 Intron NP_001129476.1
NM_015241.2 7362 UTR 3 NP_056056.2

View Full Product Details