Product Details
- SNP ID
-
hCV25995988
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:110127771 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGTTAGGGAATGAAAATTCTTTT[A/C]CAAAAGGTTTTATTGTATTGCAAGT
- Phenotype
-
MIM: 605489
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IFT81
PubMed Links
Gene Details
- Gene
- IFT81
- Gene Name
- intraflagellar transport 81
View Full Product Details