Product Details

SNP ID
rs10407417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54451122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGGGCTTCCCCTCACTCCAGC[C/G]TTTCTAGTCCACACCCAGAACTCTT
Phenotype
MIM: 616575
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LENG8 PubMed Links

Gene Details

Gene
LENG8
Gene Name
leukocyte receptor cluster member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052925.3 Intron NP_443157.1
XM_005278248.4 Intron XP_005278305.1
XM_005278249.4 Intron XP_005278306.1
XM_005278250.4 Intron XP_005278307.1
XM_005278252.4 Intron XP_005278309.1
XM_006722996.3 Intron XP_006723059.1
XM_006722997.3 Intron XP_006723060.1
XM_011526413.2 Intron XP_011524715.1
XM_011526414.2 Intron XP_011524716.1
XM_011526415.2 Intron XP_011524717.1
Gene
LENG8-AS1
Gene Name
LENG8 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details