Product Details

SNP ID
rs79312783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52783578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTAAGCAGGTGGGCTTACAGGC[A/T]ATTATTTTTTCAGAACTTTCTATAA
Phenotype
MIM: 616510 MIM: 615814
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GNPNAT1 PubMed Links

Gene Details

Gene
GNPNAT1
Gene Name
glucosamine-phosphate N-acetyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198066.3 Intron NP_932332.1
XM_005268012.2 Intron XP_005268069.1
XM_006720238.3 Intron XP_006720301.1
Gene
STYX
Gene Name
serine/threonine/tyrosine interacting protein
There are no transcripts associated with this gene.

View Full Product Details