Product Details

SNP ID
rs17157133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62297766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCCTGGGTTCTATGTTTCTATA[C/T]CATTATTGTGCTGATGACAATACCA
Phenotype
MIM: 615062
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCGB1D4 PubMed Links

Gene Details

Gene
SCGB1D4
Gene Name
secretoglobin family 1D member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206998.1 Intron NP_996881.1

View Full Product Details