Product Details

SNP ID
rs74002500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81670518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGAGGGAAGACATTGGGTTCCCA[C/T]GACGGCCCTCTGCAGGTACCCTGTT
Phenotype
MIM: 614271 MIM: 180073
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC137 PubMed Links

Gene Details

Gene
CCDC137
Gene Name
coiled-coil domain containing 137
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199287.2 Intron NP_954981.1
XM_011524738.2 Intron XP_011523040.1
XM_017024573.1 Intron XP_016880062.1
Gene
OXLD1
Gene Name
oxidoreductase like domain containing 1
There are no transcripts associated with this gene.

Gene
PDE6G
Gene Name
phosphodiesterase 6G
There are no transcripts associated with this gene.

View Full Product Details