Product Details

SNP ID
rs117923589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:26486320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCTGCTGTATACAAGGCAATAA[A/G]TGTTGTCATTATTATTGCTGAAATT
Phenotype
MIM: 606682 MIM: 602254 MIM: 612747
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HPS4 PubMed Links

Gene Details

Gene
HPS4
Gene Name
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
There are no transcripts associated with this gene.

Gene
SRRD
Gene Name
SRR1 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013694.2 Intron NP_001013716.2
XM_011530178.2 Intron XP_011528480.1
XM_017028799.1 Intron XP_016884288.1
Gene
TFIP11
Gene Name
tuftelin interacting protein 11
There are no transcripts associated with this gene.

View Full Product Details