Product Details

SNP ID
rs140200862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128713872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAAGCTTTAACTGTGAACAGCAA[C/T]GTATCAATTTATTAATTACTACGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM71F1 PubMed Links

Gene Details

Gene
FAM71F1
Gene Name
family with sequence similarity 71 member F1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282788.1 Intron NP_001269717.1
NM_001282789.1 Intron NP_001269718.1
NM_032599.3 Intron NP_115988.1
XM_017012743.1 Intron XP_016868232.1

View Full Product Details