Product Details

SNP ID
rs28715521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:67164181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTATATTATTAATGCCATGCTGC[A/G]TTTATAAGTTTGTTTTCAGCTTTTA
Phenotype
MIM: 611243
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TYW1 PubMed Links

Gene Details

Gene
TYW1
Gene Name
tRNA-yW synthesizing protein 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018264.3 Intron NP_060734.2
XM_011516372.2 Intron XP_011514674.1
XM_017012392.1 Intron XP_016867881.1

View Full Product Details