Product Details

SNP ID
rs6565529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81039304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGGCCCCAGGCACTGTCCTGCC[A/G]GTTGTTGGCTCCAGGGTGTACAACC
Phenotype
MIM: 605475
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAIAP2 PubMed Links

Gene Details

Gene
BAIAP2
Gene Name
BAI1 associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144888.1 Intron NP_001138360.1
NM_006340.2 Intron NP_006331.1
NM_017450.2 Intron NP_059344.1
NM_017451.2 Intron NP_059345.1
XM_005256943.1 Intron XP_005257000.1
XM_005256944.1 Intron XP_005257001.1
XM_005256945.1 Intron XP_005257002.1
XM_005256948.3 Intron XP_005257005.1
XM_006721635.1 Intron XP_006721698.1
XM_006721636.1 Intron XP_006721699.1
XM_006721637.2 Intron XP_006721700.1
XM_011524193.1 Intron XP_011522495.1
XM_011524194.1 Intron XP_011522496.1
XM_011524195.1 Intron XP_011522497.1
XM_011524196.1 Intron XP_011522498.1
XM_011524197.1 Intron XP_011522499.1
XM_017024017.1 Intron XP_016879506.1
XM_017024018.1 Intron XP_016879507.1
XM_017024019.1 Intron XP_016879508.1
Gene
BAIAP2-AS1
Gene Name
BAIAP2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details