Product Details

SNP ID
rs34627176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1012412 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCTGATCCCCGCGGCCCGGGACC[A/G]GGGAGGGCGGTATCTCCCAGTTCCA
Phenotype
MIM: 605830 MIM: 252800
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGFRL1 PubMed Links

Gene Details

Gene
FGFRL1
Gene Name
fibroblast growth factor receptor-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004356.2 Intron NP_001004356.1
NM_001004358.1 Intron NP_001004358.1
NM_021923.3 Intron NP_068742.2
XM_011513486.1 Intron XP_011511788.1
Gene
IDUA
Gene Name
iduronidase, alpha-L-
There are no transcripts associated with this gene.

View Full Product Details