Product Details

SNP ID
rs6418708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:17405996 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGTAGGCAGAAGCAAGAGAACCA[C/T]GTTGTGAGAGCTAGCATGGACCAAC
Phenotype
MIM: 300457
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NHS PubMed Links

Gene Details

Gene
NHS
Gene Name
NHS actin remodeling regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136024.3 Intron NP_001129496.1
NM_001291867.1 Intron NP_001278796.1
NM_001291868.1 Intron NP_001278797.1
NM_198270.3 Intron NP_938011.1
XM_011545528.2 Intron XP_011543830.1

View Full Product Details