Product Details

SNP ID
rs35937932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7678954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATCTAAAATAAAAGGCAAACAT[G/T]TTTTTTAAATGAATGTTTGCTCTCA
Phenotype
MIM: 608342
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYB5R2 PubMed Links

Gene Details

Gene
CYB5R2
Gene Name
cytochrome b5 reductase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302826.1 Intron NP_001289755.1
NM_001302827.1 Intron NP_001289756.1
NM_016229.4 Intron NP_057313.2
XM_005252975.4 Intron XP_005253032.1
XM_006718251.2 Intron XP_006718314.1
XM_011520184.2 Intron XP_011518486.1
XM_011520185.2 Intron XP_011518487.1
XM_017017920.1 Intron XP_016873409.1
XM_017017921.1 Intron XP_016873410.1
XM_017017922.1 Intron XP_016873411.1
Gene
OVCH2
Gene Name
ovochymase 2 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198185.4 Intron NP_937828.3
XM_017017659.1 Intron XP_016873148.1

View Full Product Details