Product Details

SNP ID
rs6498114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:10870261 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTACCGTAAATCTACCCCAAATAGG[G/T]TGGCAGTCTTCAAACTGAAACCAGC
Phenotype
MIM: 600005
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CIITA PubMed Links

Gene Details

Gene
CIITA
Gene Name
class II, major histocompatibility complex, transactivator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000246.3 Intron NP_000237.2
NM_001286402.1 Intron NP_001273331.1
NM_001286403.1 Intron NP_001273332.1
XM_006720880.3 Intron XP_006720943.2
XM_011522484.2 Intron XP_011520786.1
XM_011522485.2 Intron XP_011520787.1
XM_011522486.2 Intron XP_011520788.1
XM_011522487.2 Intron XP_011520789.1
XM_011522488.2 Intron XP_011520790.1
XM_011522489.2 Intron XP_011520791.1
XM_011522490.2 Intron XP_011520792.1
XM_011522491.2 Intron XP_011520793.1
XM_011522492.2 Intron XP_011520794.1
XM_011522493.2 Intron XP_011520795.1
XM_011522494.2 Intron XP_011520796.1
XM_011522495.2 Intron XP_011520797.1
XM_011522496.2 Intron XP_011520798.1

View Full Product Details