Product Details

SNP ID
rs997573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:60186062 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTGGAGTAAGAAATCACACCAGA[C/T]GGTGACTCAAATTTACAAGACAAAA
Phenotype
MIM: 114815
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CA8 PubMed Links

Gene Details

Gene
CA8
Gene Name
carbonic anhydrase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321837.1 5108 Intron NP_001308766.1
NM_001321838.1 5108 Intron NP_001308767.1
NM_001321839.1 5108 UTR 3 NP_001308768.1
NM_004056.5 5108 UTR 3 NP_004047.3
XM_011517587.2 5108 Intron XP_011515889.1
XM_011517588.2 5108 Intron XP_011515890.1
XM_017013818.1 5108 Intron XP_016869307.1

View Full Product Details