Product Details
- SNP ID
-
rs4292261
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:49111330 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAACTGGGGTGTGTGGGCATTCA[A/T]AGCTTCACTTGGATGCTGGTACCCA
- Phenotype
-
MIM: 150325
MIM: 603727
MIM: 614471
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LAMB2
PubMed Links
Gene Details
- Gene
- LAMB2
- Gene Name
- laminin subunit beta 2
There are no transcripts associated with this gene.
- Gene
- QARS
- Gene Name
- glutaminyl-tRNA synthetase
There are no transcripts associated with this gene.
- Gene
- USP19
- Gene Name
- ubiquitin specific peptidase 19
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001199160.1 |
3487 |
Missense Mutation |
ATG,TTG |
M1083L |
NP_001186089.1 |
NM_001199161.1 |
3487 |
Missense Mutation |
ATG,TTG |
M1085L |
NP_001186090.1 |
NM_001199162.1 |
3487 |
Missense Mutation |
ATG,TTG |
M1073L |
NP_001186091.1 |
NM_006677.2 |
3487 |
Missense Mutation |
ATG,TTG |
M982L |
NP_006668.1 |
XM_005264823.2 |
3487 |
Missense Mutation |
ATG,TTG |
M1087L |
XP_005264880.1 |
XM_005264824.2 |
3487 |
Missense Mutation |
ATG,TTG |
M1087L |
XP_005264881.1 |
XM_005264825.2 |
3487 |
Missense Mutation |
ATG,TTG |
M1085L |
XP_005264882.1 |
XM_005264826.2 |
3487 |
Missense Mutation |
ATG,TTG |
M1085L |
XP_005264883.1 |
XM_005264827.2 |
3487 |
Missense Mutation |
ATG,TTG |
M1072L |
XP_005264884.1 |
XM_005264829.2 |
3487 |
Missense Mutation |
ATG,TTG |
M1087L |
XP_005264886.1 |
XM_005264830.2 |
3487 |
Missense Mutation |
ATG,TTG |
M986L |
XP_005264887.1 |
XM_005264831.2 |
3487 |
Missense Mutation |
ATG,TTG |
M969L |
XP_005264888.1 |
XM_006712946.2 |
3487 |
Missense Mutation |
ATG,TTG |
M1086L |
XP_006713009.1 |
XM_006712947.2 |
3487 |
Missense Mutation |
ATG,TTG |
M1069L |
XP_006713010.1 |
XM_006712948.2 |
3487 |
Missense Mutation |
ATG,TTG |
M1083L |
XP_006713011.1 |
XM_006712949.2 |
3487 |
Missense Mutation |
ATG,TTG |
M1037L |
XP_006713012.1 |
XM_006712950.2 |
3487 |
Missense Mutation |
ATG,TTG |
M1070L |
XP_006713013.1 |
XM_006712951.2 |
3487 |
Missense Mutation |
ATG,TTG |
M1069L |
XP_006713014.1 |
XM_006712952.2 |
3487 |
Missense Mutation |
ATG,TTG |
M1068L |
XP_006713015.1 |
XM_006712953.1 |
3487 |
Missense Mutation |
ATG,TTG |
M1068L |
XP_006713016.1 |
XM_017005615.1 |
3487 |
Missense Mutation |
ATG,TTG |
M1084L |
XP_016861104.1 |
XM_017005616.1 |
3487 |
Missense Mutation |
ATG,TTG |
M1082L |
XP_016861105.1 |
XM_017005617.1 |
3487 |
Missense Mutation |
ATG,TTG |
M1070L |
XP_016861106.1 |
XM_017005618.1 |
3487 |
Missense Mutation |
ATG,TTG |
M1085L |
XP_016861107.1 |
XM_017005619.1 |
3487 |
Missense Mutation |
ATG,TTG |
M1084L |
XP_016861108.1 |
XM_017005620.1 |
3487 |
Missense Mutation |
ATG,TTG |
M1082L |
XP_016861109.1 |
XM_017005621.1 |
3487 |
Missense Mutation |
ATG,TTG |
M1035L |
XP_016861110.1 |
XM_017005622.1 |
3487 |
Missense Mutation |
ATG,TTG |
M1072L |
XP_016861111.1 |
XM_017005623.1 |
3487 |
Missense Mutation |
ATG,TTG |
M1033L |
XP_016861112.1 |
XM_017005624.1 |
3487 |
Missense Mutation |
ATG,TTG |
M1033L |
XP_016861113.1 |
XM_017005625.1 |
3487 |
Missense Mutation |
ATG,TTG |
M984L |
XP_016861114.1 |
XM_017005626.1 |
3487 |
Missense Mutation |
ATG,TTG |
M982L |
XP_016861115.1 |
XM_017005627.1 |
3487 |
Missense Mutation |
ATG,TTG |
M971L |
XP_016861116.1 |
XM_017005628.1 |
3487 |
Missense Mutation |
ATG,TTG |
M969L |
XP_016861117.1 |
XM_017005629.1 |
3487 |
Missense Mutation |
ATG,TTG |
M967L |
XP_016861118.1 |
XM_017005630.1 |
3487 |
Missense Mutation |
ATG,TTG |
M986L |
XP_016861119.1 |
XM_017005631.1 |
3487 |
Missense Mutation |
ATG,TTG |
M984L |
XP_016861120.1 |
XM_017005632.1 |
3487 |
Missense Mutation |
ATG,TTG |
M984L |
XP_016861121.1 |
XM_017005633.1 |
3487 |
Missense Mutation |
ATG,TTG |
M982L |
XP_016861122.1 |
XM_017005634.1 |
3487 |
Missense Mutation |
ATG,TTG |
M971L |
XP_016861123.1 |
XM_017005635.1 |
3487 |
Missense Mutation |
ATG,TTG |
M535L |
XP_016861124.1 |
View Full Product Details