Product Details

SNP ID
rs4405289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3228171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCCAGGGAAGCCCCGGGGTGGG[A/G]GCCGCTGGGGGCCTCGCTCCACCCG
Phenotype
MIM: 607232 MIM: 607234
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRGPRE PubMed Links

Gene Details

Gene
MRGPRE
Gene Name
MAS related GPR family member E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039165.2 936 Missense Mutation CCC,CTC P210L NP_001034254.2
Gene
MRGPRG
Gene Name
MAS related GPR family member G
There are no transcripts associated with this gene.

Gene
MRGPRG-AS1
Gene Name
MRGPRG antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details