Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_032472.3 | Intron | NP_115861.1 | |||
NM_130906.2 | Intron | NP_570981.1 | |||
XM_005246651.3 | Intron | XP_005246708.1 | |||
XM_005246652.4 | Intron | XP_005246709.1 | |||
XM_011511358.2 | Intron | XP_011509660.1 | |||
XM_017004352.1 | Intron | XP_016859841.1 | |||
XM_017004353.1 | Intron | XP_016859842.1 | |||
XM_017004354.1 | Intron | XP_016859843.1 | |||
XM_017004355.1 | Intron | XP_016859844.1 | |||
XM_017004356.1 | Intron | XP_016859845.1 | |||
XM_017004357.1 | Intron | XP_016859846.1 | |||
XM_017004358.1 | Intron | XP_016859847.1 | |||
XM_017004359.1 | Intron | XP_016859848.1 | |||
XM_017004360.1 | Intron | XP_016859849.1 |