Product Details

SNP ID
rs4254607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:28350260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTTGAGGCATCACAAGTTATTA[A/G]AAATAAAGTGGTAACCTGGTTTTTA
Phenotype
MIM: 609916
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AZI2 PubMed Links

Gene Details

Gene
AZI2
Gene Name
5-azacytidine induced 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134432.1 Intron NP_001127904.1
NM_001134433.1 Intron NP_001127905.1
NM_001271650.1 Intron NP_001258579.1
NM_022461.4 Intron NP_071906.1
Gene
ZCWPW2
Gene Name
zinc finger CW-type and PWWP domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040432.3 Intron NP_001035522.1
NM_001324169.1 Intron NP_001311098.1
NM_001324170.1 Intron NP_001311099.1

View Full Product Details