Product Details

SNP ID
rs352170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52202061 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATTAGCAGAACCTAATAAAAATA[A/G]GCTTGTGGCTGAGTGCAGTGGTTCA
Phenotype
MIM: 125290
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALAS1 PubMed Links

Gene Details

Gene
ALAS1
Gene Name
5'-aminolevulinate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000688.5 Intron NP_000679.1
NM_001304443.1 Intron NP_001291372.1
NM_001304444.1 Intron NP_001291373.1
NM_199166.2 Intron NP_954635.1
XM_011533477.1 Intron XP_011531779.1
XM_011533478.2 Intron XP_011531780.1
XM_017005873.1 Intron XP_016861362.1

View Full Product Details