Product Details
- SNP ID
-
rs2862240
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:24319556 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACCCTGTCTCAAAGCAACAAAAAAA[C/G]AAGCAGACAAACAAAACCCAGAAAA
- Phenotype
-
MIM: 614285
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C1QTNF9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76582049] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1QTNF9
- Gene Name
- C1q and tumor necrosis factor related protein 9
- Gene
- NUS1P3
- Gene Name
- NUS1 dehydrodolichyl diphosphate synthase subunit pseudogene 3
There are no transcripts associated with this gene.
View Full Product Details