Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001098491.1 | 625 | Missense Mutation | CAG,GAG | Q142E | NP_001091961.1 |
NM_001098492.1 | 625 | Missense Mutation | CAG,GAG | Q129E | NP_001091962.1 |
NM_001098493.1 | 625 | Missense Mutation | CAG,GAG | Q128E | NP_001091963.1 |
NM_001098494.1 | 625 | Missense Mutation | CAG,GAG | Q109E | NP_001091964.1 |
NM_001098495.1 | 625 | Missense Mutation | CAG,GAG | Q96E | NP_001091965.1 |
NM_001098496.1 | 625 | Missense Mutation | CAG,GAG | Q95E | NP_001091966.1 |
NM_001291743.1 | 625 | Missense Mutation | CAG,GAG | Q108E | NP_001278672.1 |
NM_001291744.1 | 625 | Missense Mutation | CAG,GAG | Q53E | NP_001278673.1 |
NM_001291745.1 | 625 | Intron | NP_001278674.1 | ||
NM_024691.3 | 625 | Missense Mutation | CAG,GAG | Q141E | NP_078967.3 |