Product Details
- SNP ID
-
rs66539868
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
26
- Location
-
Chr.1:38874330 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTACATTCGAAAGGATATCATTT[A/T]AAAAAAAAAAAATCCTGATTTGACA
- Phenotype
-
MIM: 611923
MIM: 606535
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GJA9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs535944468,rs57929942] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GJA9
- Gene Name
- gap junction protein alpha 9
There are no transcripts associated with this gene.
- Gene
- GJA9-MYCBP
- Gene Name
- GJA9-MYCBP readthrough
There are no transcripts associated with this gene.
- Gene
- LOC105378663
- Gene Name
- uncharacterized LOC105378663
There are no transcripts associated with this gene.
- Gene
- MYCBP
- Gene Name
- MYC binding protein
There are no transcripts associated with this gene.
View Full Product Details