Product Details

SNP ID
rs42650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:90349059 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACTTTCCCGCCGTTTTTGGCATA[G/T]TCACAGCCTCTTTCATGATTTCCTT
Phenotype
MIM: 610920
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GTPBP10 PubMed Links

Gene Details

Gene
GTPBP10
Gene Name
GTP binding protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042717.2 Intron NP_001036182.1
NM_033107.3 Intron NP_149098.2
Gene
LOC101927446
Gene Name
uncharacterized LOC101927446
There are no transcripts associated with this gene.

View Full Product Details