Product Details

SNP ID
rs2234964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73375880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTGCAATATTACTGTATCATTG[G/T]GATATGGCTTTACATTGATTGTATG
Phenotype
MIM: 186360
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANXA7 PubMed Links

Gene Details

Gene
ANXA7
Gene Name
annexin A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001156.4 1757 UTR 3 NP_001147.1
NM_001320879.1 1757 UTR 3 NP_001307808.1
NM_001320880.1 1757 UTR 3 NP_001307809.1
NM_004034.3 1757 UTR 3 NP_004025.1
XM_017016162.1 1757 UTR 3 XP_016871651.1
XM_017016163.1 1757 UTR 3 XP_016871652.1

View Full Product Details