Product Details

SNP ID
rs4297206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:24563181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGACCTCATACCCTCACAGATA[C/T]GAGGAGGAATCACACAATAAACAAA
Phenotype
MIM: 300948
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCYT1B PubMed Links

Gene Details

Gene
PCYT1B
Gene Name
phosphate cytidylyltransferase 1, choline, beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163264.1 Intron NP_001156736.1
NM_001163265.1 Intron NP_001156737.1
NM_004845.4 Intron NP_004836.2
XM_017029977.1 Intron XP_016885466.1

View Full Product Details