Product Details

SNP ID
rs2715275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122062842 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGATATAAGATATTTTAGGACTC[A/G]CATTCTTTGCTTTAAAATCTATTAT
Phenotype
MIM: 601020 MIM: 609739
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD86 PubMed Links

Gene Details

Gene
CD86
Gene Name
CD86 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206924.1 Intron NP_001193853.1
NM_001206925.1 Intron NP_001193854.1
NM_006889.4 Intron NP_008820.3
NM_175862.4 Intron NP_787058.4
NM_176892.1 Intron NP_795711.1
Gene
ILDR1
Gene Name
immunoglobulin like domain containing receptor 1
There are no transcripts associated with this gene.

View Full Product Details