Product Details

SNP ID
rs6444716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:193403288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCCTATATAAAGATGTGGCCCA[A/T]GTATGACATGTTTTTGATATATCCT
Phenotype
MIM: 609556
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATP13A4 PubMed Links

Gene Details

Gene
ATP13A4
Gene Name
ATPase 13A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032279.3 Intron NP_115655.2
XM_011513232.2 Intron XP_011511534.1
XM_017007318.1 Intron XP_016862807.1
XM_017007319.1 Intron XP_016862808.1

View Full Product Details