Product Details

SNP ID
rs12515443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:172196933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAAACAACAAAAAAGAAAACAA[A/C]AAAAAACTTCCCAGTTCTTTAAAGA
Phenotype
MIM: 603919
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EFCAB9 PubMed Links

Gene Details

Gene
EFCAB9
Gene Name
EF-hand calcium binding domain 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171183.1 Intron NP_001164654.1
XM_017009350.1 Intron XP_016864839.1
Gene
STK10
Gene Name
serine/threonine kinase 10
There are no transcripts associated with this gene.

View Full Product Details