Product Details
- SNP ID
-
rs12656423
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:69273432 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTGTAGCAATGCTGAAAGTAGTTT[A/G]TTCGGCTTATTTTTTCTTTCCTATA
- Phenotype
-
MIM: 613781
MIM: 601955
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC125
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112988930] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC125
- Gene Name
- coiled-coil domain containing 125
- Gene
- CDK7
- Gene Name
- cyclin dependent kinase 7
View Full Product Details