Product Details
- SNP ID
-
rs2353602
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:281327 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAGCAGCCGCATGAATCAGTTTCA[C/T]ACCCTCTCAGCTGTGATCAAGGGTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF732
PubMed Links
Gene Details
- Gene
- ZNF732
- Gene Name
- zinc finger protein 732
View Full Product Details