Product Details

SNP ID
rs12567990
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:207508340 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTAGGTTCCTCAATTGCGAAAAA[C/T]GTACCACCCTGGTGAAAGATGTTTA
Phenotype
MIM: 120620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CR1 PubMed Links

Gene Details

Gene
CR1
Gene Name
complement component 3b/4b receptor 1 (Knops blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000573.3 Intron NP_000564.2
NM_000651.4 Intron NP_000642.3
XM_006711166.2 Intron XP_006711229.1
XM_011509205.1 Intron XP_011507507.1

View Full Product Details