Product Details

SNP ID
rs2974340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42542588 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACAAGCGGTGGTCAATTTTAGAA[C/T]CACCTAGACGTGACATCAGGAACCC
Phenotype
MIM: 158378
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC20A2 PubMed Links

Gene Details

Gene
SLC20A2
Gene Name
solute carrier family 20 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257180.1 Intron NP_001244109.1
NM_001257181.1 Intron NP_001244110.1
NM_006749.4 Intron NP_006740.1
XM_005273613.3 Intron XP_005273670.1
XM_005273615.3 Intron XP_005273672.1
XM_006716390.3 Intron XP_006716453.1
XM_006716391.3 Intron XP_006716454.1
XM_017013748.1 Intron XP_016869237.1
XM_017013749.1 Intron XP_016869238.1
XM_017013750.1 Intron XP_016869239.1
XM_017013751.1 Intron XP_016869240.1
XM_017013752.1 Intron XP_016869241.1
Gene
SMIM19
Gene Name
small integral membrane protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135674.1 Intron NP_001129146.1
NM_001135675.1 Intron NP_001129147.1
NM_001135676.1 Intron NP_001129148.1
NM_138436.3 Intron NP_612445.2
XM_005273398.4 Intron XP_005273455.1

View Full Product Details