Product Details
- SNP ID
-
hCV26243314
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41609908 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAGGTGAGGCCAGCTGGTGGGCA[G/T]GAGGCTGTGGTAGAGGCAGCTCAGT
- Phenotype
-
MIM: 148067
MIM: 148069
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KRT16
PubMed Links
Gene Details
- Gene
- KRT16
- Gene Name
- keratin 16
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005557.3 |
|
Intron |
|
|
NP_005548.2 |
- Gene
- KRT17
- Gene Name
- keratin 17
There are no transcripts associated with this gene.
View Full Product Details