Product Details

SNP ID
rs1771575
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:161647707 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTTCCTTGTAAACTGGATTCCTT[C/T]GTCCTGCAGAAGAGATGTATATTGT
Phenotype
MIM: 604590
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCGR2B PubMed Links

Gene Details

Gene
FCGR2B
Gene Name
Fc fragment of IgG receptor IIb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002273.2 Intron NP_001002273.1
NM_001002274.2 Intron NP_001002274.1
NM_001002275.2 Intron NP_001002275.1
NM_001190828.1 Intron NP_001177757.1
NM_004001.4 Intron NP_003992.3
XM_011509292.2 Intron XP_011507594.1
XM_017000669.1 Intron XP_016856158.1
XM_017000670.1 Intron XP_016856159.1
XM_017000671.1 Intron XP_016856160.1
XM_017000672.1 Intron XP_016856161.1

View Full Product Details