Product Details
- SNP ID
-
rs2645992
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:112760281 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTCATTGAGCAGGAGCTGGGACCT[T/C]AGAGGAAGGGGGCCCTGGAGGGGAG
- Phenotype
-
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SNX30
PubMed Links
Gene Details
- Gene
- SNX30
- Gene Name
- sorting nexin family member 30
View Full Product Details