Product Details
- SNP ID
-
rs871788
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:112751184 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGGCGCCTGGGGCCGGGGAGTGG[G/A]AGGCTTATTTCGCTCCCCGTGGGGG
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SNX30
PubMed Links
Gene Details
- Gene
- SNX30
- Gene Name
- sorting nexin family member 30
View Full Product Details