Product Details

SNP ID
rs362150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:35066256 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAGAGGTACCCTGGGTCAGGCGC[C/T]TGTGGAGAGAGGGCTTCGCATGTAA
Phenotype
MIM: 612019
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ISX PubMed Links

Gene Details

Gene
ISX
Gene Name
intestine specific homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303508.1 121 UTR 5 NP_001290437.1

View Full Product Details