Product Details

SNP ID
hCV26322168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:19976347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGTGGTCACTTTATTCTTTGGA[C/T]CATGCATTTTCATCTATGTGTGGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4K15 PubMed Links

Gene Details

Gene
OR4K15
Gene Name
olfactory receptor family 4 subfamily K member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005486.1 829 Missense Mutation CCA,TCA P277S NP_001005486.1

View Full Product Details