Product Details

SNP ID
rs3861513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:19975867 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTCTGGTTGAGCGCAAGACTATT[G/T]CTTTTGATGCCTGCCTGGCCCAGAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4K15 PubMed Links

Gene Details

Gene
OR4K15
Gene Name
olfactory receptor family 4 subfamily K member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005486.1 349 Missense Mutation GCT,TCT A117S NP_001005486.1

View Full Product Details