Product Details

SNP ID
rs331451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:36514757 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTAATGCCACAGCTGATGTGGCAG[A/G]AGGTGGAGCTCAGGCAGTAATGTGA
Phenotype
MIM: 179615 MIM: 602355
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAG1 PubMed Links

Gene Details

Gene
RAG1
Gene Name
recombination activating gene 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000448.2 Intron NP_000439.1
XM_005253041.4 Intron XP_005253098.1
XM_011520250.2 Intron XP_011518552.1
Gene
TRAF6
Gene Name
TNF receptor associated factor 6
There are no transcripts associated with this gene.

View Full Product Details