Product Details

SNP ID
hCV26328083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18242289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGAGAACCAGAGCAACTGATCA[C/T]GTAAATCCAATTTGAGGGCAAGAGA
Phenotype
MIM: 104751 MIM: 104752
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SAA2 PubMed Links

Gene Details

Gene
SAA2
Gene Name
serum amyloid A2
There are no transcripts associated with this gene.

Gene
SAA2-SAA4
Gene Name
SAA2-SAA4 readthrough
There are no transcripts associated with this gene.

Gene
SAA4
Gene Name
serum amyloid A4, constitutive
There are no transcripts associated with this gene.

View Full Product Details