Product Details
- SNP ID
-
rs4620719
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:9282552 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTAGATGATAAAAAACAAAAATA[A/C]AATTAACAAAATAGATAACTTGATA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM41B
PubMed Links
Gene Details
- Gene
- TMEM41B
- Gene Name
- transmembrane protein 41B
View Full Product Details