Product Details

SNP ID
rs2942129
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:204400349 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTGAAAGTTAATCCATCTTTTTT[A/T]TTTTCAACATTTTTGAGACAGGGTC
Phenotype
MIM: 613257
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PPP1R15B PubMed Links

Gene Details

Gene
PPP1R15B
Gene Name
protein phosphatase 1 regulatory subunit 15B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032833.4 Intron NP_116222.4
XM_005245551.4 Intron XP_005245608.2

View Full Product Details