Product Details

SNP ID
rs34461968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109085874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTAATTTTCATTTCAGAGGTTTG[C/G]TAACAGAATGGCACTGGTATAGATT
Phenotype
MIM: 188545
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRHR PubMed Links

Gene Details

Gene
TRHR
Gene Name
thyrotropin releasing hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003301.5 130 Intron NP_003292.1
XM_011517263.2 130 UTR 5 XP_011515565.1

View Full Product Details