Product Details

SNP ID
hCV26374369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:9772524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGTTGGGTGGCCGCATTGACCC[C/T]ACCCCTGCACAATGAGAAGAGCAAG
Phenotype
MIM: 126453 MIM: 606142
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DRD5 PubMed Links

Gene Details

Gene
DRD5
Gene Name
dopamine receptor D5
There are no transcripts associated with this gene.

Gene
SLC2A9
Gene Name
solute carrier family 2 member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001290.1 Intron NP_001001290.1
NM_020041.2 Intron NP_064425.2

View Full Product Details