Product Details

SNP ID
rs2240687
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:15969155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTAAAAAACAAAAATAGACAGGA[A/G]GGGAGGGAGTCATCCTTGAATAGTG
Phenotype
MIM: 607713 MIM: 604365
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGFBP2 PubMed Links

Gene Details

Gene
FGFBP2
Gene Name
fibroblast growth factor binding protein 2
There are no transcripts associated with this gene.

Gene
PROM1
Gene Name
prominin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145847.1 3174 UTR 3 NP_001139319.1
NM_001145848.1 3174 UTR 3 NP_001139320.1
NM_001145849.1 3174 UTR 3 NP_001139321.1
NM_001145850.1 3174 UTR 3 NP_001139322.1
NM_001145851.1 3174 UTR 3 NP_001139323.1
NM_001145852.1 3174 UTR 3 NP_001139324.1
NM_006017.2 3174 UTR 3 NP_006008.1
XM_005248195.4 3174 UTR 3 XP_005248252.1
XM_005248196.4 3174 UTR 3 XP_005248253.1
XM_006713974.3 3174 UTR 3 XP_006714037.1
XM_011513890.1 3174 UTR 3 XP_011512192.1
XM_011513892.2 3174 UTR 3 XP_011512194.1
XM_011513893.2 3174 UTR 3 XP_011512195.1
XM_011513894.2 3174 UTR 3 XP_011512196.1
XM_011513895.2 3174 UTR 3 XP_011512197.1
XM_011513896.2 3174 UTR 3 XP_011512198.1
XM_011513897.2 3174 UTR 3 XP_011512199.1
XM_011513898.2 3174 Intron XP_011512200.2
XM_011513899.2 3174 Intron XP_011512201.2
XM_011513900.2 3174 UTR 3 XP_011512202.1
XM_011513902.2 3174 UTR 3 XP_011512204.1
XM_011513903.2 3174 UTR 3 XP_011512205.1
XM_017008799.1 3174 UTR 3 XP_016864288.1
XM_017008800.1 3174 Intron XP_016864289.1
XM_017008801.1 3174 Intron XP_016864290.1
XM_017008802.1 3174 UTR 3 XP_016864291.1
XM_017008803.1 3174 UTR 3 XP_016864292.1
XM_017008804.1 3174 UTR 3 XP_016864293.1
XM_017008805.1 3174 UTR 3 XP_016864294.1

View Full Product Details