Product Details

SNP ID
rs754883851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78759625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCAGAACCAGTAGCTTGGGGTA[C/T]CCAGAACCTGGCTCCCTACGCCAAC
Phenotype
MIM: 605009
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS7 PubMed Links

Gene Details

Gene
ADAMTS7
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014272.4 Intron NP_055087.2
XM_005254137.3 Intron XP_005254194.1
XM_011521166.2 Intron XP_011519468.1
Gene
LOC646938
Gene Name
TBC1 domain family member 2B pseudogene
There are no transcripts associated with this gene.

View Full Product Details