Product Details

SNP ID
rs34348079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:120690237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGGAATGAATAGATAATATATT[A/G]TTTTAACACTAATTCCAATAATGAA
Phenotype
MIM: 614161
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRDM5 PubMed Links

Gene Details

Gene
PRDM5
Gene Name
PR domain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300823.1 Intron NP_001287752.1
NM_001300824.1 Intron NP_001287753.1
NM_018699.3 Intron NP_061169.2
XM_005262708.3 Intron XP_005262765.1
XM_011531562.2 Intron XP_011529864.1
XM_011531563.2 Intron XP_011529865.1
XM_011531564.2 Intron XP_011529866.1
XM_011531565.2 Intron XP_011529867.1
XM_011531566.2 Intron XP_011529868.1
XM_011531567.2 Intron XP_011529869.1
XM_011531568.2 Intron XP_011529870.1
XM_011531569.2 Intron XP_011529871.1
XM_011531570.2 Intron XP_011529872.1
XM_017007668.1 Intron XP_016863157.1
XM_017007669.1 Intron XP_016863158.1
XM_017007670.1 Intron XP_016863159.1
XM_017007671.1 Intron XP_016863160.1

View Full Product Details