Product Details

SNP ID
rs12496318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121842653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTAATAGCTGTTATTGTTGTTTT[G/T]GTTCATATCTGTGTTCATTCTCTTA
Phenotype
MIM: 607659 MIM: 609237
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EAF2 PubMed Links

Gene Details

Gene
EAF2
Gene Name
ELL associated factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320041.1 Intron NP_001306970.1
NM_018456.5 Intron NP_060926.2
XM_005247618.3 Intron XP_005247675.1
XM_017006861.1 Intron XP_016862350.1
XM_017006862.1 Intron XP_016862351.1
XM_017006863.1 Intron XP_016862352.1
Gene
IQCB1
Gene Name
IQ motif containing B1
There are no transcripts associated with this gene.

View Full Product Details